Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006158.4 | NP_006149.2 | 1 - 2 | 1397 | 93 | |
GenBank mRNA | AK057731.1 | - | 2 - 3 | 971 | 93 | |
AK075003.1 | - | 1 - 2 | 1140 | 93 | ||
AK127430.1 | - | 3 - 4 | 840 | 93 | ||
AK225975.1 | - | 1 - 2 | 1140 | 93 | ||
AK299095.1 | - | 2 - 3 | 982 | 93 | ||
AK314591.1 | - | 1 - 2 | 1140 | 93 | ||
AL713644.1 | - | 2 - 3 | 573 | 93 | ||
AY156690.1 | - | 1 - 2 | 1043 | 93 | ||
BC039237.1 | - | 1 - 2 | 1140 | 4280159 | 93 | |
KU178228.1 | - | 2 - 3 | 764 | 93 | ||
KU178229.1 | - | 1 - 2 | 1043 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |