Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000485310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000485310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000485310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000485310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
ENST00000485310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002945.hg.1 |
TC03000165.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002945.hg.1 |
ENST00000320954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002945.hg.1 |
ENST00000423366.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002945.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002945.hg.1 |
ENST00000509775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002945.hg.1 |
ENST00000539684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002945.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148621.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148621.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148621.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148621.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148621.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300148621.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300148621.hg.1 |
TC0300006993.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000320954.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000449224.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000485310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000485310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000485310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000485310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
ENST00000485310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006993.hg.1 |
NM_006371.4 |
Refseq |