Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATAD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300946.1 | NP_001287875.1 | 9 - 10 | 1581 | 104 | |
NM_017660.3 | NP_060130.3 | 10 - 11 | 1884 | 104 | ||
XM_011528105.1 | XP_011526407.1 | 10 - 11 | 1770 | 104 | ||
XM_017026902.1 | XP_016882391.1 | 11 - 12 | 1833 | 104 | ||
XM_017026903.1 | XP_016882392.1 | 11 - 12 | 1923 | 104 | ||
XM_017026904.1 | XP_016882393.1 | 11 - 12 | 1880 | 104 | ||
XM_017026905.1 | XP_016882394.1 | 11 - 12 | 1893 | 104 | ||
XM_017026907.1 | XP_016882396.1 | 12 - 13 | 2002 | 104 | ||
XM_017026908.1 | XP_016882397.1 | 13 - 14 | 2262 | 104 | ||
XM_017026909.1 | XP_016882398.1 | 13 - 14 | 2213 | 104 | ||
XM_017026910.1 | XP_016882399.1 | 11 - 12 | 1822 | 104 | ||
XM_017026911.1 | XP_016882400.1 | 11 - 12 | 1994 | 104 | ||
GenBank mRNA | AK000092.1 | - | 2 - 3 | 326 | 104 | |
AK125974.1 | - | 12 - 13 | 1992 | 104 | ||
AK225198.1 | - | 2 - 3 | 326 | 104 | ||
AK303482.1 | - | 5 - 6 | 1092 | 104 | ||
AK315416.1 | - | 9 - 10 | 1457 | 104 | ||
AY186731.1 | - | 9 - 10 | 1572 | 104 | ||
BC011684.1 | - | 10 - 11 | 1613 | 3953055 | 104 | |
BC012902.1 | - | 3 - 4 | 472 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |