Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190458.1 | NP_001177387.1 | 41 - 42 | 5372 | 98 | |
NM_001193536.1 | NP_001180465.1 | 42 - 43 | 5468 | 98 | ||
NM_203447.3 | NP_982272.2 | 43 - 44 | 5683 | 98 | ||
XM_011518045.2 | XP_011516347.1 | 42 - 43 | 5442 | 98 | ||
XM_011518046.2 | XP_011516348.1 | 43 - 44 | 5581 | 98 | ||
XM_011518047.2 | XP_011516349.1 | 43 - 44 | 5764 | 98 | ||
XM_011518048.2 | XP_011516350.1 | 42 - 43 | 5503 | 98 | ||
XM_011518049.2 | XP_011516351.1 | 30 - 31 | 4243 | 98 | ||
XM_017015173.1 | XP_016870662.1 | 43 - 44 | 5538 | 98 | ||
XM_017015174.1 | XP_016870663.1 | 43 - 44 | 5618 | 98 | ||
GenBank mRNA | AB191037.1 | - | 43 - 44 | 5683 | 98 | |
AF194407.1 | - | 3 - 4 | 251 | 98 | ||
AK024436.1 | - | 22 - 23 | 2815 | 98 | ||
AK074081.1 | - | 29 - 30 | 3801 | 98 | ||
AK090429.1 | - | 35 - 36 | 4672 | 98 | ||
AL832270.1 | - | 41 - 42 | 7524 | 98 | ||
AY117695.1 | - | 30 - 31 | 4359 | 98 | ||
BC019102.2 | - | 8 - 9 | 1092 | 98 | ||
BC112894.1 | - | 12 - 13 | 1548 | 98 | ||
BC130518.1 | - | 42 - 43 | 5350 | 40146549 | 98 | |
BC143929.1 | - | 42 - 43 | 5350 | 9052447 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |