Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OBSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015311.2 | NP_056126.1 | 16 - 17 | 5305 | 146 | |
XM_005246424.4 | XP_005246481.1 | 15 - 16 | 5053 | 146 | ||
XM_011510857.2 | XP_011509159.1 | 16 - 17 | 5332 | 146 | ||
XM_017003696.1 | XP_016859185.1 | 16 - 17 | 5331 | 146 | ||
XM_017003697.1 | XP_016859186.1 | 16 - 17 | 5331 | 146 | ||
XM_017003698.1 | XP_016859187.1 | 15 - 16 | 5054 | 146 | ||
XM_017003699.1 | XP_016859188.1 | 15 - 16 | 5055 | 146 | ||
XM_017003700.1 | XP_016859189.1 | 14 - 15 | 4777 | 146 | ||
GenBank mRNA | EF063638.1 | - | 16 - 17 | 5048 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |