Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014581.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014581.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014581.hg.1 |
TC10001160.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000493889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000493889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000493889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000493889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
ENST00000493889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169795.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169795.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169795.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000169795.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000169795.hg.1 |
TC1000010249.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000302418.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000493889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000493889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000493889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000493889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
ENST00000493889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010249.hg.1 |
NM_004521.2 |
Refseq |