Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011528605.2 | XP_011526907.1 | 5 - 6 | 939 | 80 | |
XM_011528606.2 | XP_011526908.1 | 5 - 6 | 857 | 80 | ||
XM_011528607.2 | XP_011526909.1 | 5 - 6 | 943 | 80 | ||
XM_011528612.2 | XP_011526914.1 | 5 - 6 | 609 | 80 | ||
GenBank mRNA | AK058202.1 | - | 5 - 6 | 614 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |