Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004258.hg.1 |
ENST00000319481.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004258.hg.1 |
ENST00000357041.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004258.hg.1 |
ENST00000399443.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004258.hg.1 |
NM_001242508.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004258.hg.1 |
NM_018030.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004258.hg.1 |
NM_080597.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004258.hg.1 |
TC18000418.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
ENST00000319481.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
ENST00000357041.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
ENST00000399443.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
ENST00000578055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
ENST00000584119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
ENST00000585247.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
NM_001242508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
NM_018030.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
NM_080597.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
TC1800008301.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
ENST00000319481.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
ENST00000357041.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
ENST00000399443.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
ENST00000578055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
ENST00000584119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
ENST00000585247.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
NM_001242508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
NM_018030.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156625.hg.1 |
NM_080597.3 |
Refseq |