Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007413.hg.1 |
TC09000646.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007413.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007413.hg.1 |
ENST00000353214.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007413.hg.1 |
ENST00000465124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007413.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000465124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000465124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000465124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000465124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
ENST00000465124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008742.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007414.hg.1 |
TC09000646.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007414.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007414.hg.1 |
ENST00000353214.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007414.hg.1 |
ENST00000465124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007414.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158301.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158301.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158301.hg.1 |
ENST00000465124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158301.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158301.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158301.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158301.hg.1 |
ENST00000465124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158301.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158301.hg.1 |
TC0900008742.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000465124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000465124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000465124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000465124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
ENST00000465124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008742.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158300.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158300.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158300.hg.1 |
ENST00000465124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158300.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158300.hg.1 |
ENST00000259477.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158300.hg.1 |
ENST00000353214.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158300.hg.1 |
ENST00000465124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158300.hg.1 |
NM_030978.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158300.hg.1 |
TC0900008742.hg.1 |
Clariom D TC |