Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000474580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000474580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000474580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000474580.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000493493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000493493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000493493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000493493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008675.hg.1 |
ENST00000556041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008675.hg.1 |
ENST00000556041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008675.hg.1 |
ENST00000556041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008675.hg.1 |
ENST00000556041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008675.hg.1 |
ENST00000556041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008675.hg.1 |
ENST00000556041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008675.hg.1 |
ENST00000556041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008675.hg.1 |
ENST00000556041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300170102.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300170102.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300170102.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300170102.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300170102.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300170102.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300170102.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300170102.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300170102.hg.1 |
TC0300009696.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000474580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000474580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000474580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000474580.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000493493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000493493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000493493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000493493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |