Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008382.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008382.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008382.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008382.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008382.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008382.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008382.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008382.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008382.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008382.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008382.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008382.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008382.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008382.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008382.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008382.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008382.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008382.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006914.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006914.hg.1 |
ENST00000491766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006914.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006914.hg.1 |
TC10000629.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000156777.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156777.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156777.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156777.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156777.hg.1 |
TC1000008382.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008382.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008382.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008382.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008382.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008382.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008382.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008382.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008382.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008382.hg.1 |
ENST00000404459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008382.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008382.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008382.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008382.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008382.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008382.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008382.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008382.hg.1 |
NM_001102469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008382.hg.1 |
NM_001102469.1 |
Refseq |