Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004947.hg.1 |
TC04000362.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004947.hg.1 |
ENST00000265404.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004947.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004947.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004948.hg.1 |
TC04000362.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004948.hg.1 |
ENST00000265404.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004948.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004948.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007713.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400152595.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152595.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152595.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400152595.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152595.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152595.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400152595.hg.1 |
TC0400007713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400152596.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152596.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152596.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400152596.hg.1 |
ENST00000265404.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152596.hg.1 |
ENST00000396225.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400152596.hg.1 |
NM_012108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400152596.hg.1 |
TC0400007713.hg.1 |
Clariom D TC |