Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002821.hg.1 |
ENST00000375465.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002821.hg.1 |
ENST00000420169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002821.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002821.hg.1 |
TC20000226.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002822.hg.1 |
ENST00000375465.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002822.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002822.hg.1 |
TC20000226.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002823.hg.1 |
ENST00000375465.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002823.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002823.hg.1 |
TC20000226.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000420169.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000420169.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000420169.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000420169.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000420169.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000605211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000605211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
ENST00000605211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148959.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148959.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148959.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148959.hg.1 |
TC2000007147.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148959.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148959.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148959.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148958.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148958.hg.1 |
ENST00000420169.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148958.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148958.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148958.hg.1 |
TC2000007147.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148958.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148958.hg.1 |
ENST00000420169.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148958.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148958.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148960.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148960.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148960.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148960.hg.1 |
TC2000007147.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148960.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148960.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148960.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000375465.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000420169.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000420169.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000420169.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000420169.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000420169.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000603168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000605211.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000605211.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
ENST00000605211.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007147.hg.1 |
NR_026760.1 |
Refseq |