Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001042.hg.1 |
ENST00000298386.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001042.hg.1 |
ENST00000380314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001042.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001042.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001042.hg.1 |
TC13000106.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300146154.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146154.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146154.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146154.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146154.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146154.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146154.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146154.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146154.hg.1 |
TC1300006794.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000298386.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
ENST00000380314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_001166058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006794.hg.1 |
NM_130806.4 |
Refseq |