Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022401.hg.1 |
TC05001649.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022401.hg.1 |
ENST00000230792.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022401.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022401.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000508889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000508889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000508889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000508889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000508889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000515407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000515407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
ENST00000515407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022399.hg.1 |
TC05001649.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022399.hg.1 |
ENST00000230792.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022399.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022399.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180429.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180429.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180429.hg.1 |
ENST00000508889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180429.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180429.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180429.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180429.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180429.hg.1 |
ENST00000508889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180429.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180429.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180429.hg.1 |
TC0500011614.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500180432.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180432.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180432.hg.1 |
ENST00000508889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180432.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180432.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180432.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180432.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180432.hg.1 |
ENST00000508889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180432.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180432.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180432.hg.1 |
TC0500011614.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000230792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000507423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000508889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000508889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000508889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000508889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000508889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000515407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000515407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
ENST00000515407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_001300741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011614.hg.1 |
NM_031438.3 |
Refseq |