Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034744.hg.1 |
TC02002133.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034744.hg.1 |
ENST00000333017.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034744.hg.1 |
ENST00000409238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034744.hg.1 |
ENST00000409679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034744.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000409238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000409238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000409238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000409238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000409238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000409238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000409238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000409679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000409679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000409679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000409679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000409679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000409679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000409679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000423306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000423306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000423306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000423306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000423306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000423306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000423306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000423800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000423800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000423800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000423800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000423800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000423800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000423800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000626374.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000626374.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000626374.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000626374.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
ENST00000626374.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013605.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034747.hg.1 |
TC02002133.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034747.hg.1 |
ENST00000333017.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034747.hg.1 |
ENST00000409679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034747.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199838.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199838.hg.1 |
ENST00000409679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199838.hg.1 |
ENST00000423306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199838.hg.1 |
ENST00000626374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199838.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199838.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199838.hg.1 |
ENST00000409679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199838.hg.1 |
ENST00000423306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199838.hg.1 |
ENST00000626374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199838.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199838.hg.1 |
TC0200013605.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200199834.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199834.hg.1 |
ENST00000409238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199834.hg.1 |
ENST00000409679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199834.hg.1 |
ENST00000423306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199834.hg.1 |
ENST00000423800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199834.hg.1 |
ENST00000626374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199834.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199834.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199834.hg.1 |
ENST00000409238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199834.hg.1 |
ENST00000409679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199834.hg.1 |
ENST00000423306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199834.hg.1 |
ENST00000423800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199834.hg.1 |
ENST00000626374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199834.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199834.hg.1 |
TC0200013605.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000333017.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000409238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000409238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000409238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000409238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000409238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000409238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000409238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000409679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000409679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000409679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000409679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000409679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000409679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000409679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000423306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000423306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000423306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000423306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000423306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000423306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000423306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000423800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000423800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000423800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000423800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000423800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000423800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000423800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000626374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000626374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000626374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000626374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
ENST00000626374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
NM_175735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013605.hg.1 |
NM_175735.3 |
Refseq |