Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006710546.2 | XP_006710609.1 | 1 - 2 | 43 | 62 | |
GenBank mRNA | DA625627.1 | - | 1 - 2 | 114 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |