Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000565426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000565426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000565426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000565426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000566484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000566484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
ENST00000566484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011212.hg.1 |
ENST00000268661.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011212.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011212.hg.1 |
TC16000767.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600168842.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168842.hg.1 |
ENST00000565426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168842.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168842.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168842.hg.1 |
ENST00000565426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168842.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168842.hg.1 |
TC1600009076.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600168841.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168841.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168841.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168841.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168841.hg.1 |
TC1600009076.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000268661.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000565426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000565426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000565426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000565426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000566484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000566484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
ENST00000566484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009076.hg.1 |
NM_005061.2 |
Refseq |