Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000430354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000430354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000430354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000430354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000439107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000439107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000439107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000439107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000465574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000465574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000465574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000465574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
ENST00000465574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000707.hg.1 |
ENST00000270112.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000707.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000707.hg.1 |
TC21000113.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100146418.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146418.hg.1 |
ENST00000430354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146418.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100146418.hg.1 |
TC2100006909.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100146418.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146418.hg.1 |
ENST00000430354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146418.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000270112.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000430354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000430354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000430354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000430354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000439107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000439107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000439107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000439107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000465574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000465574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000465574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000465574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
ENST00000465574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006909.hg.1 |
NM_014586.1 |
Refseq |