Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000474665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000474665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000474665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000474665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000474665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
ENST00000474665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005055.hg.1 |
TC03000248.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005055.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005055.hg.1 |
ENST00000440097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005055.hg.1 |
ENST00000465039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005055.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005054.hg.1 |
TC03000248.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005054.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005054.hg.1 |
ENST00000440097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005054.hg.1 |
ENST00000465039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03005054.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151208.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151208.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151208.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151208.hg.1 |
ENST00000474665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151208.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151208.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151208.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151208.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151208.hg.1 |
ENST00000474665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151208.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151208.hg.1 |
TC0300007237.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300151207.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151207.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151207.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151207.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151207.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151207.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151207.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151207.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151207.hg.1 |
TC0300007237.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000273317.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000440097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000465039.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000474665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000474665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000474665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000474665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000474665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
ENST00000474665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007237.hg.1 |
NM_014240.2 |
Refseq |