Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009806.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009806.hg.1 |
ENST00000447138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009806.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009806.hg.1 |
TC20000726.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009808.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009808.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009808.hg.1 |
TC20000726.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160951.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160951.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160951.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160951.hg.1 |
TC2000008721.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160951.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160951.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160951.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160949.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160949.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160949.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160949.hg.1 |
TC2000008721.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160949.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160949.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160949.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160948.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160948.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160948.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160948.hg.1 |
TC2000008721.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160948.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160948.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160948.hg.1 |
NM_020531.2 |
Refseq |