Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024569.hg.1 |
ENST00000266743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024569.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024569.hg.1 |
ENST00000392927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024569.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024569.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024569.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024569.hg.1 |
TC12001879.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
ENST00000478238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190671.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190671.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190671.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190671.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190671.hg.1 |
ENST00000478238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190671.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190671.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190671.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190671.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190671.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190671.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190671.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190671.hg.1 |
ENST00000478238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190671.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190671.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190671.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190671.hg.1 |
TC1200011710.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000266743.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000392927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
ENST00000478238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177948.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_001177949.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011710.hg.1 |
NM_153694.4 |
Refseq |