Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010919.hg.1 |
TC03003322.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010919.hg.1 |
ENST00000264234.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010919.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010919.hg.1 |
ENST00000497685.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010919.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010920.hg.1 |
TC03003322.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010920.hg.1 |
ENST00000264234.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010920.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010920.hg.1 |
ENST00000497685.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010920.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000479520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000479520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000479520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000479520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000479520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000479520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000479520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000494855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000494855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000494855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000494855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000494855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000494855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000494855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000494855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000479520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000479520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000479520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000479520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000479520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000479520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000479520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000494855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000494855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000494855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000494855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000494855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000494855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000494855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000494855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013863.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205119.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205119.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205119.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205119.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205119.hg.1 |
ENST00000264234.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205119.hg.1 |
ENST00000460625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205119.hg.1 |
ENST00000497685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205119.hg.1 |
NM_006952.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205119.hg.1 |
TC0300013863.hg.1 |
Clariom D TC |