Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14007694.hg.1 |
ENST00000261835.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14007694.hg.1 |
ENST00000423126.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14007694.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14007694.hg.1 |
TC14000627.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000554917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000556313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000556313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000556313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000556313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000556313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000556313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000556313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000556313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000556822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000556822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000556822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
ENST00000556822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000554917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000556313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000556313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000556313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000556313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000556313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000556313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000556313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000556313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000556822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000556822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000556822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
ENST00000556822.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008182.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400158133.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400158133.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400158133.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400158133.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400158133.hg.1 |
ENST00000261835.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400158133.hg.1 |
ENST00000380228.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400158133.hg.1 |
ENST00000554611.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400158133.hg.1 |
NM_006668.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400158133.hg.1 |
TC1400008182.hg.1 |
Clariom D TC |