Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002895.hg.1 |
ENST00000229314.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002895.hg.1 |
ENST00000540141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002895.hg.1 |
ENST00000542038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002895.hg.1 |
NM_016072.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002895.hg.1 |
TC12000233.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200149469.hg.1 |
ENST00000229314.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149469.hg.1 |
ENST00000539025.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149469.hg.1 |
ENST00000539663.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149469.hg.1 |
ENST00000540141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149469.hg.1 |
ENST00000542038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149469.hg.1 |
ENST00000542194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149469.hg.1 |
NM_016072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149469.hg.1 |
ENST00000229314.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149469.hg.1 |
ENST00000539025.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149469.hg.1 |
ENST00000539663.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149469.hg.1 |
ENST00000540141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149469.hg.1 |
ENST00000542038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149469.hg.1 |
ENST00000542194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149469.hg.1 |
NM_016072.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149469.hg.1 |
TC1200007052.hg.1 |
Clariom D TC |