Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
ENST00000460409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
ENST00000460409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
ENST00000460409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010945.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021076.hg.1 |
TC07001350.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021076.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021076.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021075.hg.1 |
TC07001350.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021075.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021075.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021074.hg.1 |
TC07001350.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021074.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021074.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178247.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178247.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178247.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178247.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178247.hg.1 |
TC0700010945.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700178248.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178248.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178248.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178248.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178248.hg.1 |
TC0700010945.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700178246.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178246.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178246.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178246.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178246.hg.1 |
TC0700010945.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
ENST00000460409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
ENST00000460409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
ENST00000460409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
ENST00000490531.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
NM_001146334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010945.hg.1 |
NM_001146334.1 |
Refseq |