Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030763.2 | NP_110390.1 | 5 - 6 | 450 | 114 | |
XM_017029840.1 | XP_016885329.1 | 3 - 4 | 274 | 114 | ||
GenBank mRNA | AF250329.1 | - | 4 - 5 | 189 | 114 | |
AK094058.1 | - | 5 - 6 | 450 | 114 | ||
AK315792.1 | - | 4 - 5 | 243 | 114 | ||
BC005342.1 | - | 4 - 5 | 210 | 3838879 | 114 | |
DA745749.1 | - | 5 - 6 | 419 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons |