Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017658.hg.1 |
TC09001416.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017658.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017658.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175090.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175090.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175090.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175090.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175090.hg.1 |
TC0900011013.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
ENST00000262455.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011013.hg.1 |
NM_015051.2 |
Refseq |