Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003737.3 | NP_003728.1 | 8 - 9 | 4181 | 81 | |
GenBank mRNA | AB053446.1 | - | 8 - 9 | 4184 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |