Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000364.hg.1 |
TC6_apd_hap1000033.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000364.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000800.hg.1 |
TC6_cox_hap2000053.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000800.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000785.hg.1 |
TC6_dbb_hap3000046.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000785.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000656.hg.1 |
TC6_mcf_hap5000040.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000656.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000658.hg.1 |
TC6_mcf_hap5000040.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000658.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000794.hg.1 |
TC6_qbl_hap6000046.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000794.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000783.hg.1 |
TC6_dbb_hap3000046.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000783.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000655.hg.1 |
TC6_mcf_hap5000040.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000655.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000606.hg.1 |
TC6_ssto_hap7000041.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000606.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003394.hg.1 |
TC06000355.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003394.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003394.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003395.hg.1 |
TC06000355.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003395.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000362.hg.1 |
TC6_apd_hap1000033.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000362.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000365.hg.1 |
TC6_apd_hap1000033.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000365.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000782.hg.1 |
TC6_dbb_hap3000046.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000782.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000778.hg.1 |
TC6_mann_hap4000047.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000778.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000779.hg.1 |
TC6_mann_hap4000047.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000779.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000657.hg.1 |
TC6_mcf_hap5000040.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000657.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000608.hg.1 |
TC6_ssto_hap7000041.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000608.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000610.hg.1 |
TC6_ssto_hap7000041.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000610.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000363.hg.1 |
TC6_apd_hap1000033.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000363.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000801.hg.1 |
TC6_cox_hap2000053.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000801.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000609.hg.1 |
TC6_ssto_hap7000041.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000609.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000784.hg.1 |
TC6_dbb_hap3000046.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000784.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000777.hg.1 |
TC6_mann_hap4000047.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000777.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000796.hg.1 |
TC6_qbl_hap6000046.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000796.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000797.hg.1 |
TC6_qbl_hap6000046.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000797.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003396.hg.1 |
TC06000355.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003396.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003396.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000803.hg.1 |
TC6_cox_hap2000053.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000803.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000780.hg.1 |
TC6_mann_hap4000047.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000780.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000792.hg.1 |
TC6_qbl_hap6000046.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000792.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000607.hg.1 |
TC6_ssto_hap7000041.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000607.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000802.hg.1 |
TC6_cox_hap2000053.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000802.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000562344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000562344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000562344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000562344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000562344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000566475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000566475.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000570223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000570223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003393.hg.1 |
TC06000355.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003393.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003393.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000795.hg.1 |
TC6_qbl_hap6000046.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000795.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600151530.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151530.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151530.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151530.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600151530.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151530.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151530.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151530.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600151530.hg.1 |
TC0600007559.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600151532.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151532.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151532.hg.1 |
ENST00000566475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151532.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151532.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600151532.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151532.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151532.hg.1 |
ENST00000566475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151532.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151532.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600151532.hg.1 |
TC0600007559.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600151533.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151533.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151533.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151533.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600151533.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151533.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151533.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151533.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600151533.hg.1 |
TC0600007559.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000562344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000562344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000562344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000562344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000562344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000566475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000566475.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000570223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000570223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007559.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600151534.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151534.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151534.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151534.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600151534.hg.1 |
ENST00000426185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151534.hg.1 |
ENST00000565192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151534.hg.1 |
ENST00000615046.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151534.hg.1 |
NR_003948.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600151534.hg.1 |
TC0600007559.hg.1 |
Clariom D TC |