Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC15A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170798.1 | NP_001164269.1 | 6 - 7 | 1346 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Transporter 3' Most Homo sapiens 1700 Probe spans exons |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 25455 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P19469400 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P19469357 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P02577084 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P19469466 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P19469357 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P19469466 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P11356107 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P02577084 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3408172 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214217 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647169 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647180 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647178 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647187 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214195 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647182 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214178 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16537133 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214203 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647161 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647155 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647179 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647185 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647162 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647192 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647150 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16537138 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214212 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647189 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647164 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214166 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647160 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647194 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647193 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214191 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647158 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647183 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214193 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647166 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214197 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647156 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647151 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214171 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647164 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647183 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214212 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214178 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214197 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647162 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214203 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647169 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16537138 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647156 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647187 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647155 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647194 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214166 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214193 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214195 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647193 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647185 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647192 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647182 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16537133 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647161 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647150 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214217 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647160 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214171 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647151 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647158 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647178 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647179 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08214191 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647166 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647180 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28647189 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.1 ST Array | 7961571 | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR12016721.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12016721.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12016721.hg.1 | TC12001285.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12016719.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12016719.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12016719.hg.1 | TC12001285.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12016720.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12016720.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12016720.hg.1 | TC12001285.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200010046.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010046.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010046.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010046.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010046.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010046.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010046.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010046.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010046.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010046.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010046.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010046.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010046.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010046.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010046.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010046.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010046.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010046.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200175892.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200175892.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200175892.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200175892.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200175892.hg.1 | TC1200010046.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200175891.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200175891.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200175891.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200175891.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200175891.hg.1 | TC1200010046.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200010046.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010046.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010046.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010046.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010046.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010046.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010046.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010046.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010046.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010046.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010046.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010046.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010046.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010046.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010046.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010046.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010046.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010046.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200175890.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200175890.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200175890.hg.1 | ENST00000344941.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200175890.hg.1 | NM_001170798.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200175890.hg.1 | TC1200010046.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047382 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047382 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 107296 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1882041 | NM_001170798.1 | human_transcriptome |