Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000535820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000535820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000535820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000535820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000535820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000540893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000540893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000540893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000540893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000540893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
ENST00000540893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006974.hg.1 |
ENST00000294244.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006974.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006974.hg.1 |
TC11000576.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006975.hg.1 |
ENST00000294244.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006975.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006975.hg.1 |
TC11000576.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100156424.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156424.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156424.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100156424.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100156424.hg.1 |
TC1100007912.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000294244.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000535820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000535820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000535820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000535820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000535820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000540893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000540893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000540893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000540893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000540893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
ENST00000540893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007912.hg.1 |
NM_138471.2 |
Refseq |