Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000461652.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000461652.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000461652.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000461652.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000461652.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000461652.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
ENST00000461652.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001008.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001008.hg.1 |
ENST00000461652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001008.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001008.hg.1 |
TC13000105.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300146101.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146101.hg.1 |
ENST00000461652.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146101.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146101.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146101.hg.1 |
ENST00000461652.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146101.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146101.hg.1 |
TC1300006785.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000343307.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000461652.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000461652.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000461652.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000461652.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000461652.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000461652.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
ENST00000461652.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006785.hg.1 |
NM_194318.3 |
Refseq |