Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000558077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000558077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000558077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
ENST00000558077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15007494.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15007494.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15007494.hg.1 |
TC15000822.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500158799.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500158799.hg.1 |
ENST00000558077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500158799.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500158799.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500158799.hg.1 |
ENST00000558077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500158799.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500158799.hg.1 |
TC1500008170.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000258888.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000558077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000558077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000558077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
ENST00000558077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008170.hg.1 |
NM_020778.4 |
Refseq |