Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013436.2 | NP_001013454.1 | 1 - 2 | 132 | 65 | |
GenBank mRNA | BI463699.1 | - | 1 - 2 | 140 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |