Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015188.1 | NP_056003.1 | 11 - 12 | 2180 | 170 | |
XM_006717732.3 | XP_006717795.2 | 10 - 11 | 2099 | 170 | ||
XM_006717733.2 | XP_006717796.1 | 12 - 13 | 1140 | 170 | ||
XM_011539558.2 | XP_011537860.1 | 11 - 12 | 1191 | 170 | ||
XM_017015996.1 | XP_016871485.1 | 9 - 10 | 1955 | 170 | ||
XM_017015998.1 | XP_016871487.1 | 10 - 11 | 967 | 170 | ||
XM_017015999.1 | XP_016871488.1 | 10 - 11 | 1183 | 170 | ||
GenBank mRNA | AB011180.1 | - | 11 - 12 | 1996 | 170 | |
AB449898.1 | - | 11 - 12 | 1977 | 170 | ||
AL049956.1 | - | 10 - 11 | 928 | 170 | ||
BC064323.1 | - | 2 - 3 | 198 | 170 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |