Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000477033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000477033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000477033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000477033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
ENST00000477033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159851.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159851.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159851.hg.1 |
TC0X00008790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000477033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000477033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000477033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000477033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
ENST00000477033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008790.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159853.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159853.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159853.hg.1 |
ENST00000477033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159853.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159853.hg.1 |
ENST00000342782.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159853.hg.1 |
ENST00000370167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159853.hg.1 |
ENST00000477033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159853.hg.1 |
NM_001395.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159853.hg.1 |
TC0X00008790.hg.1 |
Clariom D TC |