Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000466.hg.1 |
TC05000039.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000466.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000466.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000513709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
ENST00000513709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000465.hg.1 |
TC05000039.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000465.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000465.hg.1 |
ENST00000536857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000465.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000511368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000513709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
ENST00000513709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006613.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500145361.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500145361.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500145361.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500145361.hg.1 |
ENST00000274181.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500145361.hg.1 |
ENST00000433402.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500145361.hg.1 |
NM_139056.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500145361.hg.1 |
TC0500006613.hg.1 |
Clariom D TC |