Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035364.hg.1 |
TC03001976.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035364.hg.1 |
ENST00000392750.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035364.hg.1 |
ENST00000461494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035364.hg.1 |
ENST00000470131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035364.hg.1 |
ENST00000471885.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035364.hg.1 |
ENST00000473645.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035364.hg.1 |
ENST00000487678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035364.hg.1 |
ENST00000487947.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035364.hg.1 |
ENST00000497056.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035364.hg.1 |
NM_007217.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035364.hg.1 |
NM_145859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035364.hg.1 |
NM_145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000392750.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000461494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000462725.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000464360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000470131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000471885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000473645.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000475915.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000481136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000483451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000487947.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000492139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000494502.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000497056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
NM_007217.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
NM_145859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
NM_145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000392750.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000461494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000462725.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000464360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000470131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000471885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000473645.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000475915.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000481136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000483451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000487947.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000492139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000494502.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
ENST00000497056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
NM_007217.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
NM_145859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
NM_145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300197585.hg.1 |
TC0300013051.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000392750.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000461494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000462725.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000464360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000470131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000471885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000473645.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000475915.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000481136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000483451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000487947.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000492139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000494502.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000497056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
NM_007217.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
NM_145859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
NM_145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000392750.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000461494.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000462725.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000464360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000470131.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000471885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000473645.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000475915.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000481136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000483451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000487947.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000492139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000494502.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
ENST00000497056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
NM_007217.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
NM_145859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
NM_145860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300197586.hg.1 |
TC0300013051.hg.1 |
Clariom D TC |