Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009411.hg.1 |
ENST00000433885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025057.hg.1 |
ENST00000301587.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025057.hg.1 |
ENST00000538432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025057.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025057.hg.1 |
TC17001869.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025058.hg.1 |
ENST00000301587.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025058.hg.1 |
ENST00000344546.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025058.hg.1 |
ENST00000538432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025058.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025058.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025058.hg.1 |
TC17001869.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025059.hg.1 |
ENST00000301587.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025059.hg.1 |
ENST00000344546.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025059.hg.1 |
ENST00000538432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025059.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025059.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025059.hg.1 |
TC17001869.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009215.hg.1 |
ENST00000567898.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025060.hg.1 |
ENST00000301587.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025060.hg.1 |
ENST00000344546.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025060.hg.1 |
ENST00000538432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025060.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025060.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025060.hg.1 |
TC17001869.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011192.hg.1 |
ENST00000414313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000344546.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000344546.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000344546.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000344546.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000344546.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000344546.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000344546.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000538432.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000538432.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000538432.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000538432.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000538432.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000538432.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000582341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000582341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
ENST00000582341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011731.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006681.hg.1 |
ENST00000396994.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000344546.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000344546.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000344546.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000344546.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000344546.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000344546.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000344546.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000538432.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000538432.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000538432.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000538432.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000538432.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000538432.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000582341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000582341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
ENST00000582341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011731.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009215.hg.1 |
ENST00000567898.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195081.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195081.hg.1 |
ENST00000344546.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195081.hg.1 |
ENST00000538432.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195081.hg.1 |
ENST00000582341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195081.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195081.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195081.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195081.hg.1 |
ENST00000344546.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195081.hg.1 |
ENST00000538432.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195081.hg.1 |
ENST00000582341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195081.hg.1 |
NM_001003785.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195081.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195081.hg.1 |
TC1700011731.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011192.hg.1 |
ENST00000414313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195078.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195078.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195078.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195078.hg.1 |
ENST00000301587.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195078.hg.1 |
ENST00000580649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195078.hg.1 |
NM_006356.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195078.hg.1 |
TC1700011731.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900006681.hg.1 |
ENST00000396994.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009411.hg.1 |
ENST00000433885.2 |
Ensembl |