Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142364.1 | NP_001135836.1 | 5 - 6 | 535 | 91 | |
NM_013338.4 | NP_037470.1 | 6 - 7 | 625 | 91 | ||
XM_017020552.1 | XP_016876041.1 | 5 - 6 | 563 | 91 | ||
GenBank mRNA | AF088028.1 | - | 1 - 2 | 63 | 91 | |
AF102850.1 | - | 6 - 7 | 633 | 91 | ||
AF161498.1 | - | 6 - 7 | 574 | 91 | ||
AF183413.1 | - | 6 - 7 | 584 | 91 | ||
AK299085.1 | - | 5 - 6 | 511 | 91 | ||
AK299126.1 | - | 6 - 7 | 594 | 91 | ||
BC012531.1 | - | 6 - 7 | 577 | 4273297 | 91 | |
BU739112.1 | - | 4 - 5 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |