Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000564236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000564236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000564236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000564236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000564236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000564236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000564333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000564333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000564333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000564333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000569093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000569093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000569093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000569093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
ENST00000569093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001418.hg.1 |
ENST00000293968.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001418.hg.1 |
ENST00000397066.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001418.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001418.hg.1 |
TC16000072.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000564236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000564236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000564236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000564236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000564236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000564236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000564333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000564333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000564333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000564333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000569093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000569093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000569093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000569093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
ENST00000569093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006610.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146604.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146604.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146604.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146604.hg.1 |
ENST00000293968.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146604.hg.1 |
ENST00000397066.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146604.hg.1 |
NM_001761.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146604.hg.1 |
TC1600006610.hg.1 |
Clariom D TC |