Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015060.2 | NP_055875.1 | 14 - 15 | 2021 | 106 | |
XM_005249668.2 | XP_005249725.1 | 14 - 15 | 1970 | 106 | ||
XM_005249670.2 | XP_005249727.1 | 14 - 15 | 1676 | 106 | ||
XM_017011891.1 | XP_016867380.1 | 14 - 15 | 1664 | 106 | ||
XM_017011892.1 | XP_016867381.1 | 14 - 15 | 1674 | 106 | ||
GenBank mRNA | AK090619.1 | - | 14 - 15 | 1883 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |