Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001326358.1 | NP_001313287.1 | 7 - 8 | 1637 | 114 | |
NM_199355.3 | NP_955387.1 | 7 - 8 | 1633 | 114 | ||
XM_011522923.1 | XP_011521225.1 | 5 - 6 | 1044 | 114 | ||
XM_011522924.1 | XP_011521226.1 | 5 - 6 | 1040 | 114 | ||
GenBank mRNA | AK131403.1 | - | 6 - 7 | 1140 | 114 | |
BC063283.1 | - | 7 - 8 | 1633 | 30343625 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Homo sapiens Probe spans exons |