Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNPY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318842.1 | NP_001305771.1 | 2 - 3 | 716 | 93 | |
NM_001318847.1 | NP_001305776.1 | 2 - 3 | 716 | 93 | ||
NR_134880.1 | 2 - 3 | 716 | 93 | |||
NR_134881.1 | 3 - 4 | 1909 | 93 | |||
GenBank mRNA | AK124196.1 | - | 3 - 4 | 1908 | 93 | |
BG675987.1 | - | 2 - 3 | 310 | 93 | ||
BQ063472.1 | - | 2 - 3 | 322 | 93 | ||
CK429647.1 | - | 2 - 3 | 329 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |