Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018420.hg.1 |
TC02001198.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018420.hg.1 |
ENST00000261016.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018420.hg.1 |
ENST00000261017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018420.hg.1 |
ENST00000261018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018420.hg.1 |
ENST00000295851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018420.hg.1 |
ENST00000422511.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018420.hg.1 |
ENST00000424558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018420.hg.1 |
ENST00000430418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018420.hg.1 |
ENST00000430574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018420.hg.1 |
NM_005759.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018419.hg.1 |
TC02001198.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018419.hg.1 |
ENST00000261016.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018419.hg.1 |
ENST00000261017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018419.hg.1 |
ENST00000261018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018419.hg.1 |
ENST00000295851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018419.hg.1 |
ENST00000422511.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018419.hg.1 |
ENST00000424558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018419.hg.1 |
ENST00000430418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018419.hg.1 |
ENST00000430574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018419.hg.1 |
NM_005759.5 |
Refseq |