Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000956.hg.1 |
TC0X002312.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000956.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000958.hg.1 |
TC0X002312.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000958.hg.1 |
ENST00000380333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000958.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000955.hg.1 |
TC0X002312.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000955.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000957.hg.1 |
TC0X002312.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000957.hg.1 |
ENST00000380333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000957.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380331.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380331.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380331.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380331.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380331.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380331.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000380336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000435806.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000435806.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000435806.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000435806.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000435806.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000435806.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000435806.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000448692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000448692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000448692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000448692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
ENST00000448692.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011276.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177243.hg.1 |
ENST00000380336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177243.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177243.hg.1 |
ENST00000380336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177243.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177243.hg.1 |
TC0X00011276.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00177249.hg.1 |
ENST00000448692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177249.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177249.hg.1 |
ENST00000448692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177249.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177249.hg.1 |
TC0X00011276.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00177250.hg.1 |
ENST00000380333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177250.hg.1 |
ENST00000448692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177250.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177250.hg.1 |
ENST00000380333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177250.hg.1 |
ENST00000448692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177250.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177250.hg.1 |
TC0X00011276.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000380336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000435806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000435806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000435806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000435806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000435806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000435806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000435806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000448692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000448692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000448692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000448692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
ENST00000448692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011276.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177254.hg.1 |
ENST00000380331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177254.hg.1 |
ENST00000380333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177254.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177254.hg.1 |
ENST00000380336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177254.hg.1 |
ENST00000435806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177254.hg.1 |
ENST00000448692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177254.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177254.hg.1 |
ENST00000380331.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177254.hg.1 |
ENST00000380333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177254.hg.1 |
ENST00000380334.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177254.hg.1 |
ENST00000380336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177254.hg.1 |
ENST00000435806.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177254.hg.1 |
ENST00000448692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177254.hg.1 |
NR_026551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177254.hg.1 |
TC0X00011276.hg.1 |
Clariom D TC |