Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUF60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271096.1 | NP_001258025.1 | 4 - 5 | 379 | 113 | |
NM_001271097.1 | NP_001258026.1 | 3 - 4 | 295 | 113 | ||
NM_001271100.1 | NP_001258029.1 | 4 - 5 | 401 | 113 | ||
NM_014281.4 | NP_055096.2 | 4 - 5 | 382 | 113 | ||
XM_011516930.2 | XP_011515232.1 | 5 - 6 | 483 | 113 | ||
XM_017013236.1 | XP_016868725.1 | 4 - 5 | 1914 | 113 | ||
XM_017013240.1 | XP_016868729.1 | 4 - 5 | 409 | 113 | ||
GenBank mRNA | AF217197.2 | - | 4 - 5 | 309 | 113 | |
BC008875.2 | - | 4 - 5 | 348 | 4138778 | 113 | |
BC009734.1 | - | 4 - 5 | 331 | 3877563 | 113 | |
BG771821.1 | - | 4 - 5 | 400 | 113 | ||
BX418925.2 | - | 4 - 5 | 331 | 113 | ||
CX163878.1 | - | 3 - 4 | 253 | 113 | ||
U51586.1 | - | 3 - 4 | 296 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |