Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | family with sequence similarity 87 member A | FAM87A | -68.7kb | Anti-sense | Chr. 8 - 375931 - 383174 on NCBI Build GRCh38 | |
Downstream | testis development related protein | TDRP |
C8orf42, Inm01, TDRP1, TDRP2 |
4.6kb | Anti-sense | Chr. 8 - 489790 - 545781 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |