Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004270.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004270.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004270.hg.1 |
TC20000302.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004271.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004271.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004271.hg.1 |
TC20000302.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007380.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000151020.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151020.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000151020.hg.1 |
TC2000007380.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000151020.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151020.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000151019.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151019.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000151019.hg.1 |
TC2000007380.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000151019.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151019.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000151022.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151022.hg.1 |
NM_003286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000151022.hg.1 |
TC2000007380.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000151022.hg.1 |
ENST00000361337.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000151022.hg.1 |
NM_003286.3 |
Refseq |